3-(4-Methoxyphenyl)propylboronsäurepinakolester ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester

H28557.06 H28557.MD H28557.03
H28557.06EA 424 EUR
H28557.06 H28557.MD H28557.03
3-(4-Methoxyphenyl)propylboronsäurepinakolester ≥97%
3-(4-Methoxyphenyl)propylboronsäurepinakolester

Purity: 97% Molecular Formula: C16H25BO3 Formula Weight: 276.19 MDL No.: MFCD09953503

Formel: C₁₆H₂₅BO₃
Molecular Weight: 276.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953503
CAS-Nummer: 1073371-72-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR