2,2-Difluor-4-pentensäure ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H32173.03 H32173.06
H32173.03EA 59.4 EUR
H32173.03 H32173.06
2,2-Difluor-4-pentensäure ≥97%
2,2-Difluoro-4-pentenoic acid

Purity: 97% CAS No.: 55039-89-9 Molecular Formula: C5H6F2O2 Formula Weight: 136.10 Boiling Point: 56-58°/3mm UN No.: 3265 MDL No.: MFCD09800642

Formel: C₅H₆F₂O₂
Siedepunkt: 56…58 °C (3 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD09800642
CAS-Nummer: 55039-89-9
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR