4-Nitrophenylboronsäurepinakolester ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester
H52848.03
H52848.06
H52848.03EA
31.6
EUR
H52848.03
H52848.06
4-Nitrophenylboronsäurepinakolester ≥98%
4-Nitrophenylboronsäurepinakolester
Purity: 98%
CAS No.: 171364-83-3
Molecular Formula: C12H16BNO4
Formula Weight: 249.08
Melting Point: 108-109°
MDL No.: MFCD02179437
Formel:
C₁₂H₁₆BNO₄ Molecular Weight: 249.07 g/mol Schmelzpunkt: 108…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179437 CAS-Nummer: 171364-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...