2-Phenoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Phenoxybenzeneboronic acid
H53264.03
H53264.06
H53264.03EA
36.3
EUR
H53264.03
H53264.06
2-Phenoxyphenylboronsäure ≥98%
2-Phenoxyphenylboronsäure
Purity: 98%
CAS No.: 108238-09-1
Molecular Formula: C12H11BO3
Formula Weight: 214.03
Melting Point: 116°
MDL No.: MFCD01001592
Formel:
C₁₂H₁₁BO₃ Molecular Weight: 214,03 g/mol Schmelzpunkt: 116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01001592 CAS-Nummer: 108238-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...