1-Benzyloxy-3-iodbenzol ≥99%

Lieferant: Thermo Scientific

Synonyme: 1-(Benzyloxy)-3-iodobenzene

L17442.03 L17442.06
L17442.03EA 12.2 EUR
L17442.03 L17442.06
1-Benzyloxy-3-iodbenzol ≥99%
1-Benzyloxy-3-iodbenzol

Purity: 99% CAS No.: 107623-21-2 Molecular Formula: C13H11IO Formula Weight: 310.14 Melting Point: 50-51° Beilstein Reference: 3270112

Formel: C₁₃H₁₁IO
Molecular Weight: 310,13 g/mol
Schmelzpunkt: 50…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318100
CAS-Nummer: 107623-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR