n-Dodecan-[D26] (98% D)
Lieferant: LGC Standards PROMOCHEM
CIL-DLM-338-1
CIL-DLM-338-5
PROOCIL-DLM-338-1EA
247
EUR
PROOCIL-DLM-338-1
PROOCIL-DLM-338-5
n-Dodecan-[D26] (98% D)
n-Dodecan-[D26]
Formel:
D₃C(CD₂)₁₀CD₃ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
16416-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...