2-Fluor-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2258-1G BB-2258-250M
COBBBB-2258-1GEA 709 EUR
COBBBB-2258-1G COBBBB-2258-250M
2-Fluor-3-nitrophenylboronsäure
2-Fluor-3-nitrophenylboronsäure
Formel: C₆H₅BFNO₄
MDL: MFCD12025960
CAS-Nummer: 1150114-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR