2-Phenoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Phenoxybenzeneboronic acid

BB-2459-100G BB-2459-5G BB-2459-1G BB-2459-25G
COBBBB-2459-100GEA 0 EUR
COBBBB-2459-100G COBBBB-2459-5G COBBBB-2459-1G COBBBB-2459-25G
2-Phenoxyphenylboronsäure
2-Phenoxyphenylboronsäure
Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
MDL: MFCD01001592
CAS-Nummer: 108238-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR