2,4-Dibrom-5-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2593-5G BB-2593-1G
COBBBB-2593-5GEA 0 EUR
COBBBB-2593-5G COBBBB-2593-1G
2,4-Dibrom-5-methoxyphenylboronsäure
2,4-Dibrom-5-methoxyphenylboronsäure
Formel: C₇H₇BBr₂O₃
Molecular Weight: 309.75 g/mol
MDL: MFCD06659860
CAS-Nummer: 89677-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR