2,6-Dibrom-3-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2594-1G
BB-2594-5G
COBBBB-2594-1GEA
257
EUR
COBBBB-2594-1G
COBBBB-2594-5G
2,6-Dibrom-3-methoxyphenylboronsäure
2,6-Dibrom-3-methoxyphenylboronsäure
Formel:
C₇H₇BBr₂O₃ |
MDL:
MFCD06659861 CAS-Nummer: 850567-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...