2,4-Dichlor-5-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2,4-Dichloro-5-methoxyphenyl)boronic acid
BB-2598-1G
BB-2598-5G
BB-2598-250M
COBBBB-2598-1GEA
692
EUR
COBBBB-2598-1G
COBBBB-2598-5G
COBBBB-2598-250M
2,4-Dichlor-5-methoxyphenylboronsäure
2,4-Dichlor-5-methoxyphenylboronsäure
Formel:
C₇H₇BCl₂O₃ Molecular Weight: 220.85 g/mol |
MDL:
MFCD12546454 CAS-Nummer: 431942-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...