3,5-Dimethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dimethoxybenzeneboronic acid

BB-2622-5G BB-2622-25G BB-2622-1G
COBBBB-2622-5GEA 63.2 EUR
COBBBB-2622-5G COBBBB-2622-25G COBBBB-2622-1G
3,5-Dimethoxyphenylboronsäure
3,5-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181,98 g/mol
MDL: MFCD03095127
CAS-Nummer: 192182-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR