2,3,6-Trimethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,3,6-Trimethoxybenzeneboronic acid, (2,3,6-Trimethoxyphenyl)boronic acid

BB-2634-5G BB-2634-1G
COBBBB-2634-5GEA 620 EUR
COBBBB-2634-5G COBBBB-2634-1G
2,3,6-Trimethoxyphenylboronsäure
2,3,6-Trimethoxyphenylboronsäure
Formel: C₉H₁₃BO₅
Molecular Weight: 212,01 g/mol
MDL: MFCD03411944
CAS-Nummer: 380430-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR