2-Chlor-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3470-1G BB-3470-5G
COBBBB-3470-1GEA 58.5 EUR
COBBBB-3470-1G COBBBB-3470-5G
2-Chlor-5-nitrophenylboronsäure
2-Chlor-5-nitrophenylboronsäure
Formel: C₆H₅BClNO₄
Molecular Weight: 201,37 g/mol
MDL: MFCD03095323
CAS-Nummer: 867333-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR