2,3-Difluor-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,3-Difluoro-4-methoxybenzeneboronic acid, (2,3-Difluoro-4-methoxyphenyl)boronic acid

BB-3808-5G BB-3808-25G BB-3808-1G
COBBBB-3808-5GEA 95.9 EUR
COBBBB-3808-5G COBBBB-3808-25G COBBBB-3808-1G
2,3-Difluor-4-methoxyphenylboronsäure
2,3-Difluor-4-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
Molecular Weight: 187,94 g/mol
MDL: MFCD07368234
CAS-Nummer: 170981-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR