2,3-Difluor-6-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3809-1G BB-3809-25G BB-3809-5G
COBBBB-3809-1GEA 0 EUR
COBBBB-3809-1G COBBBB-3809-25G COBBBB-3809-5G
2,3-Difluor-6-methoxyphenylboronsäure
2,3-Difluor-6-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
Molecular Weight: 187.94 g/mol
MDL: MFCD03095116
CAS-Nummer: 957061-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR