2,6-Difluor-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3810-1G BB-3810-5G BB-3810-25G
COBBBB-3810-1GEA 38.2 EUR
COBBBB-3810-1G COBBBB-3810-5G COBBBB-3810-25G
2,6-Difluor-3-methoxyphenylboronsäure
2,6-Difluor-3-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
Molecular Weight: 187,94 g/mol
MDL: MFCD06657880
CAS-Nummer: 870779-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR