2,6-Difluor-3-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2,6-Difluoro-3-hydroxyphenyl)boronic acid

BB-5095-1G BB-5095-5G
COBBBB-5095-1GEA 210 EUR
COBBBB-5095-1G COBBBB-5095-5G
2,6-Difluor-3-hydroxyphenylboronsäure
2,6-Difluor-3-hydroxyphenylboronsäure
Formel: C₆H₅BF₂O₃
Molecular Weight: 173,91 g/mol
MDL: MFCD09878345
CAS-Nummer: 957065-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR