2,4-Dichlor-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5219-5G BB-5219-1G
COBBBB-5219-5GEA 1540 EUR
COBBBB-5219-5G COBBBB-5219-1G
2,4-Dichlor-3-methoxyphenylboronsäure
2,4-Dichlor-3-methoxyphenylboronsäure
Formel: C₇H₇BCl₂O₃
MDL: MFCD10699638
CAS-Nummer: 944128-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR