2-Fluor-3-isopropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluoro-3-isopropoxyphenyl)boronic acid

BB-5281-5G BB-5281-1G
COBBBB-5281-5GEA 1490 EUR
COBBBB-5281-5G COBBBB-5281-1G
2-Fluor-3-isopropoxyphenylboronsäure
2-Fluor-3-isopropoxyphenylboronsäure
Formel: C₉H₁₂BFO₃
Molecular Weight: 198 g/mol
MDL: MFCD11504844
CAS-Nummer: 855230-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR