2,5-Dibrom-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-6091-5G BB-6091-1G BB-6091-250M
COBBBB-6091-5GEA 2040 EUR
COBBBB-6091-5G COBBBB-6091-1G COBBBB-6091-250M
2,5-Dibrom-4-methoxyphenylboronsäure
2,5-Dibrom-4-methoxyphenylboronsäure
Formel: C₇H₇BBr₂O₃
Molecular Weight: 309.75 g/mol
MDL: MFCD13195657
CAS-Nummer: 1217501-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR