2,4-Dibrom-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8016-5G BB-8016-1G
COBBBB-8016-5GEA 204 EUR
COBBBB-8016-5G COBBBB-8016-1G
2,4-Dibrom-6-fluorphenylboronsäure
2,4-Dibrom-6-fluorphenylboronsäure
Formel: C₆H₄BBr₂FO₂
Molecular Weight: 297.71 g/mol
MDL: MFCD07784352
CAS-Nummer: 870778-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR