2,6-Difluor-3-propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8064-1G BB-8064-250M
COBBBB-8064-1GEA 0 EUR
COBBBB-8064-1G COBBBB-8064-250M
2,6-Difluor-3-propoxyphenylboronsäure
2,6-Difluor-3-propoxyphenylboronsäure
Formel: C₉H₁₁BF₂O₃
MDL: MFCD07369752
CAS-Nummer: 849062-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR