2-Brom-4,5-difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8111-1G BB-8111-250M
COBBBB-8111-1GEA 175 EUR
COBBBB-8111-1G COBBBB-8111-250M
2-Brom-4,5-difluorphenylboronsäure
2-Brom-4,5-difluorphenylboronsäure
Formel: C₆H₄BBrF₂O₂
MDL: MFCD06411353
CAS-Nummer: 849062-34-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR