2-Brom-3-ethoxy-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-3-ethoxy-6-fluorophenyl)boronic acid

BB-8169-1G BB-8169-250M
COBBBB-8169-1GEA 266 EUR
COBBBB-8169-1G COBBBB-8169-250M
2-Brom-3-ethoxy-6-fluorphenylboronsäure
2-Brom-3-ethoxy-6-fluorphenylboronsäure
Formel: C₈H₉BBrFO₃
Molecular Weight: 262,87 g/mol
MDL: MFCD06411368
CAS-Nummer: 849052-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR