2-Brom-3-butoxy-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-3-butoxy-6-fluorophenyl)boronic acid

BB-8206-1G BB-8206-250M
COBBBB-8206-1GEA 257 EUR
COBBBB-8206-1G COBBBB-8206-250M
2-Brom-3-butoxy-6-fluorphenylboronsäure
2-Brom-3-butoxy-6-fluorphenylboronsäure
Formel: C₁₀H₁₃BBrFO₃
Molecular Weight: 290,92 g/mol
MDL: MFCD08276824
CAS-Nummer: 1072951-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR