2,4-Dipropoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2,4-Dipropoxyphenyl)boronic acid

BB-8211-1G BB-8211-250M
COBBBB-8211-1GEA 330 EUR
COBBBB-8211-1G COBBBB-8211-250M
2,4-Dipropoxyphenylboronsäure
2,4-Dipropoxyphenylboronsäure
Formel: C₁₂H₁₉BO₄
Molecular Weight: 238.09 g/mol
MDL: MFCD09265142
CAS-Nummer: 150145-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR