2-Amino-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8405-1G BB-8405-250M
COBBBB-8405-1GEA 1870 EUR
COBBBB-8405-1G COBBBB-8405-250M
2-Amino-5-fluorphenylboronsäure
2-Amino-5-fluorphenylboronsäure
Formel: C₆H₇BFNO₂
Molecular Weight: 154.94 g/mol
MDL: MFCD03095362
CAS-Nummer: 1040400-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR