2,5-Difluor-4-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2,5-Difluoro-4-methoxyphenyl)boronic acid
BB-8451-1G
BB-8451-5G
COBBBB-8451-1GEA
313
EUR
COBBBB-8451-1G
COBBBB-8451-5G
2,5-Difluor-4-methoxyphenylboronsäure
2,5-Difluor-4-methoxyphenylboronsäure
Formel:
C₇H₇BF₂O₃ Molecular Weight: 187.94 g/mol |
MDL:
MFCD04038754 CAS-Nummer: 897958-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...