2-Brom-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-5-fluorophenyl)boronic acid

BB-8566-1G BB-8566-5G
COBBBB-8566-1GEA 175 EUR
COBBBB-8566-1G COBBBB-8566-5G
2-Brom-5-fluorphenylboronsäure
2-Brom-5-fluorphenylboronsäure
Formel: C₆H₅BBrFO₂
Molecular Weight: 218,82 g/mol
MDL: MFCD08690232
CAS-Nummer: 959996-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR