2,3-Difluor-4-hexyloxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2,3-Difluoro-4-(n-hexyloxy)phenylboronic acid
BB-9012-5G
BB-9012-1G
COBBBB-9012-5GEA
0
EUR
COBBBB-9012-5G
COBBBB-9012-1G
2,3-Difluor-4-hexyloxyphenylboronsäure
2,3-Difluor-4-hexyloxyphenylboronsäure
Formel:
C₁₂H₁₇BF₂O₃ |
MDL:
MFCD06656273 CAS-Nummer: 121219-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...