2-Hydroxy-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

FM-2438-1G FM-2438-250M
COBBFM-2438-1GEA 930 EUR
COBBFM-2438-1G COBBFM-2438-250M
2-Hydroxy-4-pyridinboronsäurepinakolester
2-Hydroxy-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221.06 g/mol
MDL: MFCD11223287
CAS-Nummer: 1256358-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR