2-Benzyloxy-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

FM-3254-1G FM-3254-250M
COBBFM-3254-1GEA 1110 EUR
COBBFM-3254-1G COBBFM-3254-250M
2-Benzyloxy-4-pyridinboronsäurepinakolester
2-Benzyloxy-4-pyridinboronsäurepinakolester
Formel: C₁₈H₂₂BNO₃
MDL: MFCD11878170
CAS-Nummer: 1256359-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR