4-Methylphenylboronsäureneopentylglycolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methylphenylboronic acid neopentyl ester, 5,5-Dimethyl-2-(p-tolyl)-1,3,2-dioxaborinane, 4-Methylbenzeneboronic acid neopentyl glycol ester

NE-2337-5G NE-2337-25G NE-2337-1G
COBBNE-2337-5GEA 186 EUR
COBBNE-2337-5G COBBNE-2337-25G COBBNE-2337-1G
4-Methylphenylboronsäureneopentylglycolester
4-Methylphenylboronsäureneopentylglycolester
Formel: C₁₂H₁₇BO₂
Molecular Weight: 204,08 g/mol
MDL: MFCD03788728
CAS-Nummer: 380481-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR