4-Bromphenylboronsäureneopentylglycolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 4-Bromobenzeneboronic acid neopentyl glycol ester

NE-2629-1G NE-2629-5G
COBBNE-2629-1GEA 63.4 EUR
COBBNE-2629-1G COBBNE-2629-5G
4-Bromphenylboronsäureneopentylglycolester
4-Bromphenylboronsäureneopentylglycolester
Formel: C₁₁H₁₄BBrO₂
Molecular Weight: 268,94 g/mol
MDL: MFCD01318105
CAS-Nummer: 183677-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR