1-Benzyloxy-3-fluorbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluorobenzyloxybenzene

OT-0503-5G OT-0503-25G OT-0503-1G
COBBOT-0503-5GEA 314 EUR
COBBOT-0503-5G COBBOT-0503-25G COBBOT-0503-1G
1-Benzyloxy-3-fluorbenzol
1-Benzyloxy-3-fluorbenzol
Formel: C₁₃H₁₁FO
Molecular Weight: 202,23 g/mol
MDL: MFCD06409983
CAS-Nummer: 72216-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR