2-Cyan-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

PN-0143-1G PN-0143-250M
COBBPN-0143-1GEA 911 EUR
COBBPN-0143-1G COBBPN-0143-250M
2-Cyan-4-pyridinboronsäurepinakolester
2-Cyan-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230,07 g/mol
MDL: MFCD09607734
CAS-Nummer: 741709-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR