2-Pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-pyridine

PN-2096-1G PN-2096-5G
COBBPN-2096-1GEA 276 EUR
COBBPN-2096-1G COBBPN-2096-5G
2-Pyridinboronsäurepinakolester
2-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205,06 g/mol
MDL: MFCD04039342
CAS-Nummer: 874186-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR