2-Chlor-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-3-boronic acid pinacol ester

PN-2100-5G PN-2100-1G
COBBPN-2100-5GEA 101 EUR
COBBPN-2100-5G COBBPN-2100-1G
2-Chlor-3-pyridinboronsäurepinakolester
2-Chlor-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239.51 g/mol
MDL: MFCD04039873
CAS-Nummer: 452972-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR