4-Cyanphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester

PN-2264-5G PN-2264-25G PN-2264-1G
COBBPN-2264-5GEA 125 EUR
COBBPN-2264-5G COBBPN-2264-25G COBBPN-2264-1G
4-Cyanphenylboronsäurepinakolester
4-Cyanphenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₂
Molecular Weight: 229,09 g/mol
MDL: MFCD03093897
CAS-Nummer: 171364-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR