2-Ethoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Ethoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Ethoxypyridine-3-boronic acid pinacol ester, 2-Ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-2271-5G PN-2271-1G
COBBPN-2271-5GEA 484 EUR
COBBPN-2271-5G COBBPN-2271-1G
2-Ethoxy-3-pyridinboronsäurepinakolester
2-Ethoxy-3-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₃
Molecular Weight: 249.12 g/mol
MDL: MFCD07366721
CAS-Nummer: 848243-23-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR