2-Chlor-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chloropyridine-4-boronic acid pinacol ester, 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-4-boronic acid Pinacol ester
PN-2272-5G
PN-2272-1G
COBBPN-2272-5GEA
312
EUR
COBBPN-2272-5G
COBBPN-2272-1G
2-Chlor-4-pyridinboronsäurepinakolester
2-Chlor-4-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BClNO₂ Molecular Weight: 239,51 g/mol |
MDL:
MFCD04039870 CAS-Nummer: 458532-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...