2,6-Difluor-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Difluoropyridine-3-boronic acid, pinacol ester, 2,6-Difluoropyridine-3-boronic acid Pinacol ester, 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2,6-Difluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2275-1G PN-2275-5G
COBBPN-2275-1GEA 196 EUR
COBBPN-2275-1G COBBPN-2275-5G
2,6-Difluor-3-pyridinboronsäurepinakolester
2,6-Difluor-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₄BF₂NO₂
Molecular Weight: 241,04 g/mol
MDL: MFCD09037475
CAS-Nummer: 1072945-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR