2-Ethoxy-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Ethoxypyridine-5-boronic acid pinacol ester, 2-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-2278-5G
PN-2278-1G
COBBPN-2278-5GEA
497
EUR
COBBPN-2278-5G
COBBPN-2278-1G
2-Ethoxy-5-pyridinboronsäurepinakolester
2-Ethoxy-5-pyridinboronsäurepinakolester
Formel:
C₁₃H₂₀BNO₃ Molecular Weight: 249.12 g/mol |
MDL:
MFCD07781182 CAS-Nummer: 1072945-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...