2-Methoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methoxybenzeneboronic acid pinacol ester, 2-Methoxyphenylboronic acid, pinacol ester, 2-(2-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2324-5G
PN-2324-25G
PN-2324-1G
COBBPN-2324-5GEA
166
EUR
COBBPN-2324-5G
COBBPN-2324-25G
COBBPN-2324-1G
2-Methoxyphenylboronsäurepinakolester
2-Methoxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₉BO₃ Molecular Weight: 234,10 g/mol |
MDL:
MFCD05863916 CAS-Nummer: 190788-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...