4-Methoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methoxyphenylboronic acid Pinacol ester

PN-2326-5G PN-2326-1G
COBBPN-2326-5GEA 130 EUR
COBBPN-2326-5G COBBPN-2326-1G
4-Methoxyphenylboronsäurepinakolester
4-Methoxyphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₃
Molecular Weight: 234,1 g/mol
MDL: MFCD05663864
CAS-Nummer: 171364-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR