2-Methoxy-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methoxypyridine-3-boronic acid, pinacol ester, 2-Methoxypyridine-5-boronic acid, pinacol ester

PN-2329-5G PN-2329-1G
COBBPN-2329-5GEA 127 EUR
COBBPN-2329-5G COBBPN-2329-1G
2-Methoxy-5-pyridinboronsäurepinakolester
2-Methoxy-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235,09 g/mol
MDL: MFCD05663858
CAS-Nummer: 445264-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR