2-Methylphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4,4,5,5-Tetramethyl-2-(o-tolyl)-1,3,2-dioxaborolane, 2-Methylphenylboronic acid, pinacol ester, o-Tolylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene
PN-2335-5G
PN-2335-1G
COBBPN-2335-5GEA
235
EUR
COBBPN-2335-5G
COBBPN-2335-1G
2-Methylphenylboronsäurepinakolester
2-Methylphenylboronsäurepinakolester
Formel:
C₁₃H₁₉BO₂ Molecular Weight: 218.1 g/mol |
MDL:
MFCD06795658 CAS-Nummer: 195062-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...