3-Methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methylbenzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane, 3-Tolylboronic acid pinacol ester, m-Tolylboronic acid Pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene

PN-2336-5G PN-2336-25G PN-2336-1G
COBBPN-2336-5GEA 76.1 EUR
COBBPN-2336-5G COBBPN-2336-25G COBBPN-2336-1G
3-Methylphenylboronsäurepinakolester
3-Methylphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₂
Molecular Weight: 218.1 g/mol
MDL: MFCD05864303
CAS-Nummer: 253342-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR