4-Methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane

PN-2337-100G PN-2337-5G PN-2337-1G PN-2337-25G
COBBPN-2337-100GEA 548 EUR
COBBPN-2337-100G COBBPN-2337-5G COBBPN-2337-1G COBBPN-2337-25G
4-Methylphenylboronsäurepinakolester
4-Methylphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₂
Molecular Weight: 218.1 g/mol
MDL: MFCD03453662
CAS-Nummer: 195062-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR